Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLMAP SIRNA Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.3: 57756221 - 57930013 on Build GRCh38
UniGene ID:
Hs.476432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19189
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304420.2
NP_001291349.1
14
1769
NM_001304421.2
NP_001291350.1
12
1655
NM_001304422.2
NP_001291351.1
2
251
NM_001304423.2
NP_001291352.1
2
251
NM_001311178.1
NP_001298107.1
2
251
NM_001311179.1
NP_001298108.1
2
251
NM_007159.4
NP_009090.2
13
1718
XM_005265456.3
XP_005265513.1
16
2656
XM_005265457.3
XP_005265514.1
16
2650
XM_005265458.3
XP_005265515.1
16
2656
XM_005265459.3
XP_005265516.1
15
2605
XM_005265460.3
XP_005265517.1
15
2605
XM_005265461.3
XP_005265518.1
15
2599
XM_005265462.3
XP_005265519.1
15
2593
XM_005265464.3
XP_005265521.1
15
2587
XM_005265465.3
XP_005265522.1
14
2554
XM_005265466.3
XP_005265523.1
14
2548
XM_005265467.3
XP_005265524.1
14
2542
XM_005265468.3
XP_005265525.1
16
2656
XM_005265470.3
XP_005265527.1
13
2491
XM_005265471.3
XP_005265528.1
13
2485
XM_005265472.3
XP_005265529.1
14
2542
XM_006713326.2
XP_006713389.1
14
2548
XM_011534103.1
XP_011532405.1
14
2542
XM_011534104.1
XP_011532406.1
14
2554
XM_011534105.1
XP_011532407.1
13
2491
XM_011534106.1
XP_011532408.1
2
126
XM_017007146.1
XP_016862635.1
16
2650
XM_017007147.1
XP_016862636.1
15
2599
XM_017007148.1
XP_016862637.1
15
2605
XM_017007149.1
XP_016862638.1
15
2599
XM_017007150.1
XP_016862639.1
15
2599
XM_017007151.1
XP_016862640.1
14
2554
XM_017007152.1
XP_016862641.1
14
2548
XM_017007153.1
XP_016862642.1
14
2536
XM_017007154.1
XP_016862643.1
14
2536
XM_017007155.1
XP_016862644.1
16
2650
XM_017007156.1
XP_016862645.1
16
2656
XM_017007157.1
XP_016862646.1
16
2650
XM_017007158.1
XP_016862647.1
13
2491
XM_017007159.1
XP_016862648.1
15
2605
XM_017007160.1
XP_016862649.1
13
2485
XM_017007161.1
XP_016862650.1
15
2599
XM_017007162.1
XP_016862651.1
15
2605
XM_017007163.1
XP_016862652.1
15
2599
XM_017007164.1
XP_016862653.1
14
2548
XM_017007165.1
XP_016862654.1
14
2536
XM_017007166.1
XP_016862655.1
14
2536
XM_017007167.1
XP_016862656.1
13
2491
XM_017007168.1
XP_016862657.1
13
2485
XM_017007169.1
XP_016862658.1
16
2656
XM_017007170.1
XP_016862659.1
2
126
GenBank mRNA
AB046821.1
-
11
959
AF100750.1
-
2
195
AF304450.1
-
2
124
AK124200.1
-
14
1459
AK300106.1
-
2
174
AK302934.1
-
2
126
AK304493.1
-
2
178
AK316436.1
-
2
119
BC114627.1
-
12
1353
40081892
BC115701.1
-
14
1467
40081893
CR627321.1
-
2
108
DB100357.1
-
2
255
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0