Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Safb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022394.1 | NP_071789.1 | 4, 5 | 638 | ||
GenBank mRNA | AF056324.1 | - | 4, 5 | 638 |