Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033870.1 | NP_001029042.1 | 5 | 873 | ||
NM_023102.2 | NP_075590.2 | 5 | 870 | |||
XM_006241000.2 | XP_006241062.1 | 4 | 2248 | |||
XM_006241001.2 | XP_006241063.1 | 4 | 956 | |||
XM_006241002.2 | XP_006241064.1 | 5 | 2168 | |||
XM_006241003.2 | XP_006241065.1 | 4 | 1598 | |||
XM_008765130.1 | XP_008763352.1 | 4 | 956 | |||
GenBank mRNA | BC072533.1 | - | 5 | 873 | 7098992 | |
U22297.1 | - | 4 | 604 |