Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 12, 13 | 2316 | ||
XM_005263862.3 | XP_005263919.1 | 12, 13 | 2605 | |||
XM_005263863.4 | XP_005263920.1 | 12, 13 | 2392 | |||
XM_005263864.3 | XP_005263921.1 | 12, 13 | 2557 | |||
XM_011510507.1 | XP_011508809.1 | 14, 15 | 2447 | |||
XM_017003198.1 | XP_016858687.1 | 11, 12 | 2512 | |||
XM_017003199.1 | XP_016858688.1 | 11, 12 | 2465 | |||
GenBank mRNA | AB017114.1 | - | 8, 9 | 1444 | ||
AB449889.1 | - | 12, 13 | 2185 | |||
AK292826.1 | - | 10, 11 | 1949 | |||
BQ673509.1 | - | 3, 4 | 534 |