Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031035.3 | NP_112297.1 | 9 | 1684 | ||
XM_006243858.1 | XP_006243920.1 | 9 | 1327 | |||
GenBank mRNA | BC078806.1 | - | 9 | 1636 | 7121183 | |
M12672.1 | - | 9 | 1581 | |||
M17528.1 | - | 9 | 1683 |