Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031069.1 | NP_112331.1 | 5 | 648 | ||
GenBank mRNA | U48246.1 | - | 5 | 648 |