Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031982.1 | NP_114188.1 | 3, 4 | 525 | ||
XM_008767882.1 | XP_008766104.1 | 2, 3 | 532 | |||
GenBank mRNA | AB040873.1 | - | 2, 3 | 445 | ||
AB041029.1 | - | 2, 3 | 445 | |||
AF029310.1 | - | 3, 4 | 525 |