Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053421.1 | NP_445873.1 | 4 | 236 | ||
XM_006254016.2 | XP_006254078.1 | 5 | 561 | |||
XM_006254019.2 | XP_006254081.1 | 5 | 561 | |||
XM_006254020.2 | XP_006254082.1 | 5 | 561 | |||
XM_008771764.1 | XP_008769986.1 | 5 | 561 | |||
XM_008771765.1 | XP_008769987.1 | 5 | 561 | |||
XM_008771766.1 | XP_008769988.1 | 5 | 561 | |||
GenBank mRNA | AF245512.1 | - | 4 | 236 |
Gene Symbol: LOC100912163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771836.1 | XP_008770058.1 | 4 | 321 |