Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318891.1 | NP_001305820.1 | 6 | 1032 | ||
NM_001318892.1 | NP_001305821.1 | 6 | 880 | |||
NM_001318893.1 | NP_001305822.1 | 6 | 1061 | |||
NM_007149.2 | NP_009080.2 | 6 | 788 | |||
XM_011514861.2 | XP_011513163.1 | 6 | 814 | |||
GenBank mRNA | AK123011.1 | - | 5 | 1314 | ||
AK131264.1 | - | 6 | 1040 | |||
AK312804.1 | - | 6 | 1029 | |||
BC022992.1 | - | 6 | 770 | 4997766 | ||
U66561.1 | - | 3 | 428 |