Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053531.1 | NP_445983.1 | 2 | 179 | ||
XM_006249016.2 | XP_006249078.1 | 3 | 332 | |||
GenBank mRNA | AF281632.1 | - | 2 | 179 | ||
FQ228923.1 | - | 3 | 292 | |||
FQ230649.1 | - | 2 | 103 |