Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145047.1 | NP_001138519.1 | 3 | 171 | ||
NM_001145048.1 | NP_001138520.1 | 2 | 103 | |||
NM_001145049.1 | NP_001138521.1 | 2 | 103 | |||
NM_007266.3 | NP_009197.2 | 2 | 210 | |||
NR_026735.1 | 2 | 210 | ||||
GenBank mRNA | AB044661.1 | - | 2 | 171 | ||
AJ010842.1 | - | 2 | 100 | |||
AK298827.1 | - | 2 | 504 | |||
AK298866.1 | - | 2 | 210 | |||
AK302131.1 | - | 2 | 176 | |||
AK310340.1 | - | 3 | 161 | |||
AK311423.1 | - | 2 | 233 | |||
AK312992.1 | - | 2 | 182 | |||
AK316404.1 | - | 2 | 103 | |||
BC007451.2 | - | 2 | 154 | 3029376 |