Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133591.2 | NP_598275.1 | 9 | 1560 | ||
XM_008767941.1 | XP_008766163.1 | 9 | 1918 | |||
XM_008767942.1 | XP_008766164.1 | 9 | 1983 | |||
GenBank mRNA | AF022774.1 | - | 8 | 1501 | ||
BC091126.1 | - | 9 | 1560 | 7380477 |