Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138535.2 | NP_612544.2 | 24 | 4334 | ||
XM_006236895.2 | XP_006236957.1 | 23 | 5211 | |||
GenBank mRNA | AF090113.1 | - | 4573 | |||
AF205193.1 | - | 24 | 4336 |