Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138858.1 | NP_620213.1 | 2 | 278 | ||
GenBank mRNA | AF100172.1 | - | 2 | 278 |