Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138913.1 | NP_620268.1 | 2 | 392 | ||
XM_006249382.2 | XP_006249444.1 | 2 | 436 | |||
GenBank mRNA | AY221507.1 | - | 2 | 408 | ||
BC086979.1 | - | 2 | 379 | 7191865 | ||
FQ231421.1 | - | 2 | 421 | |||
FQ231433.1 | - | 2 | 397 | |||
FQ232784.1 | - | 2 | 422 | |||
Z18877.1 | - | 2 | 410 |