Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139042.2 | NP_620611.2 | 1, 2 | 393 | ||
GenBank mRNA | AF024622.1 | - | 1, 2 | 393 |