Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139337.1 | NP_647553.1 | 2 | 131 | ||
XM_006230677.2 | XP_006230739.1 | 2 | 465 | |||
GenBank mRNA | AF404762.1 | - | 2 | 131 | ||
AF419856.1 | - | 2 | 164 | |||
FQ216840.1 | - | 2 | 298 |