Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012079.5 | NP_036211.2 | 2 | 507 | ||
XM_011517356.2 | XP_011515658.1 | 2 | 228 | |||
GenBank mRNA | AB057815.1 | - | 2 | 250 | ||
AF059202.1 | - | 2 | 468 | |||
BC015762.1 | - | 2 | 472 | 4863905 | ||
BC023565.1 | - | 2 | 477 | 4130484 | ||
BC150649.1 | - | 2 | 453 | 9057020 | ||
HY016503.1 | - | 2 | 507 |