Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007202.3 | NP_009133.2 | 3 | 445 | ||
XM_006721432.3 | XP_006721495.1 | 3 | 440 | |||
XM_017024071.1 | XP_016879560.1 | 3 | 440 | |||
GenBank mRNA | AF037439.1 | - | 3 | 405 | ||
AK295180.1 | - | 3 | 386 | |||
AK308297.1 | - | 3 | 292 | |||
AK312438.1 | - | 3 | 398 | |||
BC017055.1 | - | 3 | 401 | 3882941 | ||
BP229845.1 | - | 3 | 445 | |||
DQ270159.1 | - | 3 | 268 | |||
KU178546.1 | - | 3 | 268 | |||
KU178547.1 | - | 3 | 268 |