Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153302.1 | NP_695214.1 | 6 | 1136 | ||
GenBank mRNA | AF411249.1 | - | 6 | 1136 | ||
BC089773.1 | - | 6 | 1131 | |||
BC105900.1 | - | 6 | 1141 | 7377732 |