Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007218.3 | NP_009149.2 | 2 | 1029 | ||
XM_017013002.1 | XP_016868491.1 | 3 | 719 | |||
GenBank mRNA | AF064801.1 | - | 2 | 923 | ||
AK001602.1 | - | 2 | 732 | |||
AK025043.1 | - | 1 | 238 | |||
AK098519.1 | - | 1 | 103 | |||
BC021571.1 | - | 2 | 900 | 4550103 | ||
BC064636.1 | - | 2 | 896 | 5748185 |