Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021776.1 | NP_068544.1 | 1 | 545 | ||
XM_006245475.1 | XP_006245537.1 | 2 | 653 | |||
GenBank mRNA | AB023896.1 | - | 1 | 502 | ||
AB026293.1 | - | 1 | 545 |