Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022618.1 | NP_072140.1 | 2 | 288 | ||
GenBank mRNA | AF139518.1 | - | 2 | 288 |