Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234591.2 | XP_006234653.1 | 5 | 814 | ||
XM_006234592.2 | XP_006234654.1 | 5 | 762 | |||
XM_006234593.2 | XP_006234655.1 | 5 | 1439 | |||
GenBank mRNA | AF315950.1 | - | 5 | 728 | ||
BC157805.1 | - | 5 | 680 | 9026085 |