Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031005.3 | NP_112267.1 | 17 | 2309 | ||
GenBank mRNA | AF115386.1 | - | 17 | 2309 | ||
AY437436.1 | - | 17 | 2100 | |||
BC074001.1 | - | 2353 | 7099133 |