Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277211.1 | NP_001264140.1 | 11 | 1080 | ||
NM_001277212.1 | NP_001264141.1 | 11 | 1068 | |||
NM_001277213.1 | NP_001264142.1 | 11 | 1065 | |||
NM_031352.2 | NP_112642.1 | 11 | 1071 | |||
GenBank mRNA | AB009346.1 | - | 11 | 1051 | ||
AB038364.1 | - | 11 | 994 | |||
AB038365.1 | - | 11 | 1051 | |||
AB039818.1 | - | 11 | 1008 | |||
AB039819.1 | - | 11 | 1039 | |||
BC072483.1 | - | 11 | 1057 | 7104722 | ||
FQ228854.1 | - | 11 | 1068 |