Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031662.1 | NP_113850.1 | 16 | 1806 | ||
XM_006246799.2 | XP_006246861.1 | 17 | 2006 | |||
XM_006246801.2 | XP_006246863.1 | 17 | 1953 | |||
XM_008767866.1 | XP_008766088.1 | 18 | 2095 | |||
GenBank mRNA | AB023658.1 | - | 16 | 1806 | ||
L42810.1 | - | 16 | 1806 | |||
S83194.1 | - | 16 | 1806 |