Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123377.1 | NP_001116849.1 | 895 | |||
NM_007262.4 | NP_009193.2 | 953 | ||||
GenBank mRNA | D61380.2 | - | 796 | |||
EU794641.1 | - | 796 |