Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145047.1 | NP_001138519.1 | 15 | 1396 | ||
NM_001145048.1 | NP_001138520.1 | 14 | 1328 | |||
NM_001145049.1 | NP_001138521.1 | 12 | 1221 | |||
NM_007266.3 | NP_009197.2 | 14 | 1435 | |||
NR_026735.1 | 12 | 1328 | ||||
GenBank mRNA | AB044661.1 | - | 14 | 1396 | ||
AJ010842.1 | - | 14 | 1325 | |||
AK302131.1 | - | 12 | 1294 | |||
BC007451.2 | - | 14 | 1379 | 3029376 |