Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031834.1 | NP_114022.1 | 8 | 989 | ||
GenBank mRNA | FQ209630.1 | - | 8 | 998 | ||
FQ210886.1 | - | 8 | 994 | |||
FQ214244.1 | - | 8 | 994 | |||
FQ218459.1 | - | 8 | 994 | |||
FQ218506.1 | - | 8 | 1004 | |||
FQ218658.1 | - | 8 | 993 | |||
FQ219178.1 | - | 8 | 993 | |||
FQ219521.1 | - | 8 | 994 | |||
FQ219542.1 | - | 8 | 994 | |||
FQ219948.1 | - | 8 | 994 | |||
L19998.1 | - | 8 | 989 | |||
X52883.1 | - | 8 | 977 | |||
X68640.1 | - | 7 | 894 |