Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007282.4 | NP_009213.1 | 7 | 1206 | ||
NM_183381.2 | NP_899237.1 | 6 | 1104 | |||
XM_005247092.3 | XP_005247149.1 | 6 | 1063 | |||
XM_011512373.2 | XP_011510675.1 | 8 | 1211 | |||
XM_011512374.2 | XP_011510676.1 | 7 | 1109 | |||
XM_011512376.2 | XP_011510678.1 | 6 | 514 | |||
XM_017005654.1 | XP_016861143.1 | 7 | 1285 | |||
XM_017005655.1 | XP_016861144.1 | 6 | 1186 | |||
XM_017005656.1 | XP_016861145.1 | 7 | 1165 | |||
XM_017005657.1 | XP_016861146.1 | 7 | 1298 | |||
XM_017005658.1 | XP_016861147.1 | 6 | 1196 | |||
XM_017005659.1 | XP_016861148.1 | 5 | 933 | |||
XM_017005660.1 | XP_016861149.1 | 5 | 923 | |||
GenBank mRNA | AF037204.1 | - | 6 | 573 | ||
AF070558.1 | - | 6 | 564 | |||
AK090638.1 | - | 6 | 1084 | |||
AK090771.1 | - | 3 | 193 | |||
AK313304.1 | - | 6 | 588 | |||
BC009781.1 | - | 6 | 581 | 3683407 | ||
BC009803.1 | - | 7 | 684 | 4274992 | ||
BC017878.1 | - | 6 | 546 | |||
BG546092.1 | - | 4 | 377 | |||
BM556246.1 | - | 5 | 459 | |||
CR456804.1 | - | 5 | 422 |