Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032080.1 | NP_114469.1 | 2 | 303 | ||
XM_006248373.2 | XP_006248435.1 | 2 | 1009 | |||
XM_006248374.2 | XP_006248436.1 | 2 | 1009 | |||
XM_006248375.2 | XP_006248437.1 | 2 | 1009 | |||
XM_008768745.1 | XP_008766967.1 | 2 | 1009 | |||
GenBank mRNA | X53428.1 | - | 2 | 278 | ||
X73653.1 | - | 2 | 303 |