Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053348.1 | NP_445800.1 | 1 | 314 | ||
XM_006248549.1 | XP_006248611.1 | 2 | 268 | |||
XM_006248550.1 | XP_006248612.1 | 2 | 631 | |||
GenBank mRNA | AJ242926.1 | - | 1 | 314 | ||
BC070508.1 | - | 2 | 253 | 7099154 | ||
FQ209398.1 | - | 1 | 325 | |||
FQ209449.1 | - | 2 | 249 | |||
FQ209518.1 | - | 1 | 265 | |||
FQ209610.1 | - | 1 | 300 | |||
FQ209832.1 | - | 1 | 265 | |||
FQ210294.1 | - | 1 | 265 | |||
FQ210335.1 | - | 1 | 266 | |||
FQ211050.1 | - | 1 | 265 | |||
FQ211248.1 | - | 1 | 300 | |||
FQ218476.1 | - | 1 | 296 | |||
FQ218557.1 | - | 2 | 244 | |||
FQ218709.1 | - | 1 | 265 | |||
FQ218884.1 | - | 2 | 236 | |||
FQ218893.1 | - | 1 | 265 | |||
FQ218905.1 | - | 1 | 265 | |||
FQ218930.1 | - | 1 | 298 | |||
FQ218949.1 | - | 1 | 115 | |||
FQ219048.1 | - | 1 | 296 | |||
FQ219152.1 | - | 1 | 302 | |||
FQ219170.1 | - | 1 | 265 | |||
FQ219190.1 | - | 1 | 300 | |||
FQ219266.1 | - | 1 | 266 | |||
FQ219352.1 | - | 1 | 265 | |||
FQ219454.1 | - | 1 | 265 | |||
FQ219459.1 | - | 1 | 263 | |||
FQ219500.1 | - | 1 | 268 | |||
FQ219513.1 | - | 1 | 337 |