Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 18 | 2263 | ||
XM_006256924.2 | XP_006256986.2 | 18 | 2205 | |||
XM_006256929.2 | XP_006256991.2 | 16 | 2031 | |||
XM_006256930.2 | XP_006256992.1 | 13 | 1624 | |||
XM_006256931.1 | XP_006256993.1 | 20 | 2556 | |||
XM_008773199.1 | XP_008771421.1 | 7 | 814 | |||
XM_008773200.1 | XP_008771422.1 | 7 | 754 | |||
XM_008773201.1 | XP_008771423.1 | 7 | 710 | |||
XM_008773202.1 | XP_008771424.1 | 6 | 606 | |||
GenBank mRNA | AF131867.1 | - | 16 | 1819 | ||
AF230520.1 | - | 18 | 1929 | |||
AF255884.1 | - | 6 | 446 | |||
AF255885.1 | - | 6 | 981 | |||
AF255886.1 | - | 14 | 1500 | |||
AF255887.1 | - | 13 | 1394 | |||
AF255888.1 | - | 18 | 2181 | |||
BC070877.1 | - | 18 | 2263 | 7096160 | ||
FQ226336.1 | - | 9 | 951 | |||
U90261.2 | - | 2 | 71 |