Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007361.3 | NP_031387.3 | 21, 22 | 4359 | ||
XM_005267405.4 | XP_005267462.1 | 20, 21 | 4443 | |||
XM_005267406.4 | XP_005267463.1 | 19, 20 | 4299 | |||
XM_005267407.4 | XP_005267464.1 | 20, 21 | 4218 | |||
GenBank mRNA | AJ223500.1 | - | 21, 22 | 4125 | ||
AK291652.1 | - | 21, 22 | 4187 | |||
AK298542.1 | - | 12, 13 | 1930 | |||
AK300462.1 | - | 21, 22 | 3853 | |||
BC035608.1 | - | 17, 18 | 2961 | 5205293 | ||
BX648241.1 | - | 22, 23 | 5533 | |||
D86425.1 | - | 21, 22 | 4128 |