Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053471.2 | NP_445923.1 | 6 | 709 | ||
XM_006244348.2 | XP_006244410.1 | 7 | 1178 | |||
XM_006244349.2 | XP_006244411.1 | 6 | 531 | |||
GenBank mRNA | AB000776.1 | - | 6 | 709 |