Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207048.1 | NP_001193977.1 | 2 | 209 | ||
NM_001207049.1 | NP_001193978.1 | 2 | 209 | |||
NM_012104.4 | NP_036236.1 | 3 | 847 | |||
NM_138971.3 | NP_620427.1 | 3 | 847 | |||
NM_138972.3 | NP_620428.1 | 3 | 847 | |||
NM_138973.3 | NP_620429.1 | 3 | 847 | |||
GenBank mRNA | AB032975.2 | - | 3 | 829 | ||
AB050436.1 | - | 3 | 389 | |||
AB050437.1 | - | 3 | 389 | |||
AB050438.1 | - | 3 | 389 | |||
AB177847.1 | - | 2 | 71 | |||
AF190725.1 | - | 3 | 839 | |||
AF200193.1 | - | 3 | 347 | |||
AF200343.1 | - | 3 | 386 | |||
AF201468.1 | - | 3 | 840 | |||
AF204943.1 | - | 3 | 386 | |||
AF338816.1 | - | 3 | 386 | |||
AF338817.1 | - | 3 | 386 | |||
AK075049.1 | - | 3 | 322 | |||
AK296015.1 | - | 2 | 112 | |||
AK296535.1 | - | 2 | 122 | |||
BC036084.1 | - | 3 | 842 | 5311572 | ||
BC065492.1 | - | 3 | 847 | 6538154 | ||
DC343475.1 | - | 2 | 209 |