Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130428.1 | NP_569112.1 | 3, 4 | 332 | ||
GenBank mRNA | AB072907.1 | - | 3, 4 | 332 | ||
DQ402976.1 | - | 1, 2 | 141 |