Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133526.1 | NP_598210.1 | 8 | 1115 | ||
GenBank mRNA | BC061785.1 | - | 8 | 971 | 5623788 | |
FQ214867.1 | - | 8 | 1024 | |||
FQ215876.1 | - | 4 | 487 | |||
FQ216712.1 | - | 8 | 1144 | |||
FQ220073.1 | - | 4 | 487 | |||
FQ220093.1 | - | 4 | 487 | |||
FQ227464.1 | - | 8 | 1132 | |||
FQ229225.1 | - | 4 | 487 | |||
FQ229522.1 | - | 8 | 1033 | |||
Y13275.2 | - | 8 | 1115 |