Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134365.2 | NP_599192.1 | 7 | 951 | ||
GenBank mRNA | BC063808.1 | - | 7 | 892 | 6920021 | |
FQ209675.1 | - | 7 | 850 | |||
FQ212544.1 | - | 7 | 887 | |||
FQ214336.1 | - | 7 | 879 | |||
FQ214353.1 | - | 7 | 913 | |||
FQ214371.1 | - | 7 | 887 | |||
FQ214449.1 | - | 7 | 850 | |||
FQ214672.1 | - | 7 | 886 | |||
FQ214784.1 | - | 7 | 885 | |||
FQ215244.1 | - | 7 | 850 | |||
FQ215436.1 | - | 7 | 850 | |||
FQ215480.1 | - | 7 | 850 | |||
FQ215788.1 | - | 7 | 886 | |||
FQ215823.1 | - | 7 | 903 | |||
FQ216062.1 | - | 7 | 853 | |||
FQ216247.1 | - | 7 | 836 | |||
FQ216773.1 | - | 7 | 850 | |||
FQ216829.1 | - | 7 | 850 | |||
FQ216922.1 | - | 7 | 850 | |||
FQ218701.1 | - | 7 | 886 | |||
FQ219899.1 | - | 7 | 880 | |||
FQ223652.1 | - | 7 | 850 | |||
FQ223683.1 | - | 7 | 876 | |||
FQ229261.1 | - | 7 | 910 | |||
FQ232356.1 | - | 7 | 887 | |||
M35052.1 | - | 7 | 870 |
Gene Symbol: LOC100911417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749372.2 | XP_003749420.1 | 946 |