Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145879.2 | NP_665886.2 | 1 | 514 | ||
GenBank mRNA | AW917497.1 | - | 2 | 515 |