Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178093.2 | NP_835194.2 | 2 | 434 | ||
XM_006253180.2 | XP_006253242.1 | 6 | 3049 | |||
XM_006253181.2 | XP_006253243.1 | 6 | 2749 | |||
XM_006253182.2 | XP_006253244.1 | 4 | 1369 | |||
XM_008771280.1 | XP_008769502.1 | 3 | 793 | |||
GenBank mRNA | AB190507.1 | - | 2 | 511 | ||
AY208915.1 | - | 2 | 202 | |||
BC072537.1 | - | 2 | 434 | |||
BC166591.1 | - | 2 | 483 | 7938925 | ||
EU417559.1 | - | 5 | 2552 | |||
GU245886.1 | - | 4 | 442 |