Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284501.1 | NP_001271430.1 | 3, 4 | 431 | ||
NM_012225.3 | NP_036357.1 | 2, 3 | 241 | |||
XM_005255027.2 | XP_005255084.1 | 2, 3 | 708 | |||
XM_017022831.1 | XP_016878320.1 | 2, 3 | 375 | |||
GenBank mRNA | AF118394.1 | - | 2, 3 | 206 | ||
AK001023.1 | - | 2, 3 | 217 | |||
AK315611.1 | - | 2, 3 | 182 | |||
BC002768.2 | - | 2, 3 | 198 | 3633393 | ||
BC008005.1 | - | 2, 3 | 223 | 3507768 | ||
BU189242.1 | - | 2, 3 | 241 |