Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181381.2 | NP_852046.1 | 5 | 564 | ||
XM_006236572.2 | XP_006236634.1 | 5 | 601 | |||
XM_006236573.2 | XP_006236635.1 | 5 | 735 | |||
XM_006236574.2 | XP_006236636.1 | 5 | 655 | |||
XM_006236575.2 | XP_006236637.1 | 5 | 1255 | |||
XM_006236576.1 | XP_006236638.1 | 5 | 666 | |||
XM_006236577.2 | XP_006236639.1 | 4 | 566 | |||
XM_008762961.1 | XP_008761183.1 | 5 | 625 | |||
GenBank mRNA | AB094089.1 | - | 4 | 483 | ||
AB105817.1 | - | 5 | 538 | |||
AY089996.1 | - | 5 | 564 | |||
AY089997.1 | - | 5 | 564 | |||
AY089998.1 | - | 5 | 564 |