Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111294.1 | NP_001104764.1 | 2 | 166 | ||
NM_001111295.1 | NP_001104765.1 | 2 | 100 | |||
NM_198132.3 | NP_937765.1 | 2 | 166 | |||
XM_008761911.1 | XP_008760133.1 | 2 | 115 | |||
XM_008761912.1 | XP_008760134.1 | 2 | 115 | |||
XM_008761913.1 | XP_008760135.1 | 2 | 161 | |||
GenBank mRNA | AY363226.1 | - | 2 | 112 | ||
AY363227.1 | - | 2 | 46 | |||
BC081878.1 | - | 2 | 106 | 7108128 | ||
CB585714.1 | - | 2 | 178 | |||
CB725005.1 | - | 2 | 38 | |||
FQ214617.1 | - | 2 | 62 |