Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198762.1 | NP_942057.1 | 9 | 711 | ||
XM_006249942.2 | XP_006250004.1 | 9 | 1035 | |||
XM_006249943.2 | XP_006250005.1 | 9 | 1035 | |||
XM_008769564.1 | XP_008767786.1 | 10 | 1110 | |||
GenBank mRNA | AY359443.1 | - | 9 | 711 |