Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198773.1 | NP_942068.1 | 7 | 1908 | ||
GenBank mRNA | AB081729.1 | - | 7 | 1908 |