Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080153.1 | NP_001073622.1 | 3 | 591 | ||
NM_198781.2 | NP_942076.2 | 3 | 501 | |||
XM_006235944.2 | XP_006236006.1 | 3 | 1237 | |||
XM_006235945.2 | XP_006236007.1 | 3 | 628 | |||
XM_008762651.1 | XP_008760873.1 | 3 | 503 | |||
GenBank mRNA | AY603378.1 | - | 3 | 591 | ||
DY313123.1 | - | 3 | 501 |