Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012134.2 | NP_036266.2 | 1 | 478 | ||
GenBank mRNA | AK304852.1 | - | 1 | 397 | ||
AK311740.1 | - | 1 | 406 | |||
BC001755.1 | - | 1 | 359 | |||
BC080187.1 | - | 1 | 427 | 6177902 | ||
DB228115.1 | - | 1 | 478 | |||
X54162.1 | - | 1 | 358 |