Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271319.1 | NP_001258248.1 | 3 | 511 | ||
NM_199393.2 | NP_955425.2 | 2 | 428 | |||
XM_006235922.2 | XP_006235984.1 | 1 | 569 | |||
XM_006235923.2 | XP_006235985.1 | 2 | 921 | |||
GenBank mRNA | BC062004.1 | - | 2 | 362 | 5599809 | |
FQ223626.1 | - | 3 | 511 |